accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3SOOOO

CAS RN 33963-55-2

IUPAC Name: 2-methanesulfonylbenzoate
Synonyms: 2-methanesulfonylbenzoate
Molecular Weight (g/mol): 199.2
Molecular Formula: C8H7O4S
InChi Key: BZSXEZOLBIJVQK-UHFFFAOYSA-M
SMILES: CS(=O)(=O)C1=CC=CC=C1C([O-])=O
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)