accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2NNH2NClN

CAS RN 3397-62-4

IUPAC Name: 6-chloro-1,3,5-triazine-2,4-diamine
Synonyms: 2-chloro-4,6-diaminotriazine
Molecular Weight (g/mol): 145.55
Molecular Formula: C3H4ClN5
InChi Key: FVFVNNKYKYZTJU-UHFFFAOYSA-N
SMILES: NC1=NC(Cl)=NC(N)=N1
Concentration 
  • (3)
Solvent or Matrix 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results