accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHNHOHO

CAS RN 3398-22-9

IUPAC Name: 4-hydroxypyrrolidine-2-carboxylic acid
Synonyms: hydroxyproline 4Hyp 4-hydroxy-L-prolines 4-hydroxyproline
Molecular Weight (g/mol): 131.13
Molecular Formula: C5H9NO3
InChi Key: PMMYEEVYMWASQN-UHFFFAOYNA-N
SMILES: OC1CNC(C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)