accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 33996-33-7

H3CON(S)OH(S)OO

CAS RN 33996-33-7

IUPAC Name: (2S,4S)-1-acetyl-4-hydroxypyrrolidine-2-carboxylate
Synonyms: acetyl hydroxyproline
Molecular Weight (g/mol): 172.16
Molecular Formula: C7H10NO4
InChi Key: BAPRUDZDYCKSOQ-WDSKDSINSA-M
SMILES: CC(=O)N1C[C@@H](O)C[C@H]1C([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)