CAS RN 34054-55-2
CAS RN 34054-55-2
IUPAC Name:
yttrium(3+) hydrate phosphate
Synonyms:
yttrium(3+) hydrate phosphate
Molecular Weight (g/mol):
201.89
Molecular Formula:
H2O5PY
InChi Key:
ICIYNWVOHBLOFM-UHFFFAOYSA-K
SMILES:
O.[Y+3].[O-]P([O-])([O-])=O
1
–
1
of
1
results
1
Yttrium(III) phosphate hydrate, 99.99% (REO)
CAS: 34054-55-2 Molecular Formula: O4PY Molecular Weight (g/mol): 183.88 MDL Number: MFCD00049782 InChI Key: UXBZSSBXGPYSIL-UHFFFAOYSA-K Synonym: yttrium iii phosphate hydrate,acmc-20aj99,yttrium phosphate-water 1/1/1,yttrium 3+ hydrate phosphate,yttrium 3+ ion hydrate phosphate PubChem CID: 57372096 IUPAC Name: yttrium(3+) phosphate SMILES: [Y+3].[O-]P([O-])([O-])=O