accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HOOONOO

CAS RN 34071-95-9

IUPAC Name: 2,5-dioxopyrrolidin-1-yl 3-(4-hydroxyphenyl)propanoate
Synonyms: 2,5-dioxopyrrolidin-1-yl 3-(4-hydroxyphenyl)propanoate
Molecular Weight (g/mol): 263.25
Molecular Formula: C13H13NO5
InChi Key: KYRUKRFVOACELK-UHFFFAOYSA-N
SMILES: OC1=CC=C(CCC(=O)ON2C(=O)CCC2=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results