accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 34091-51-5

IUPAC Name: 5-iodo-1-methyl-1H-pyrazole
Synonyms: 5-iodo-1-methylpyrazole
Molecular Weight (g/mol): 208
Molecular Formula: C4H5IN2
InChi Key: RJYWUQWCLZYCTI-UHFFFAOYSA-N
SMILES: CN1N=CC=C1I

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results