accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 3411-03-8

CH3OCl

CAS RN 3411-03-8

IUPAC Name: 3-chloro-4-methylbenzaldehyde
Synonyms: 3-chloro-4-methylbenzaldehyde
Molecular Weight (g/mol): 154.59
Molecular Formula: C8H7ClO
InChi Key: OJFQNFVCICBYQC-UHFFFAOYSA-N
SMILES: CC1=C(Cl)C=C(C=O)C=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)