accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 3417-91-2

HONH2(S)OOCH3HCl

CAS RN 3417-91-2

IUPAC Name: methyl 2-amino-3-(4-hydroxyphenyl)propanoate hydrochloridyl
Synonyms: methyl tyrosinate hcl
Molecular Weight (g/mol): 230.67
Molecular Formula: C10H13ClNO3
InChi Key: XPXBJIXLKIUUKZ-UHFFFAOYNA-N
SMILES: [Cl].COC(=O)C(N)CC1=CC=C(O)C=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)