accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3(S)(S)PNH2

CAS RN 341968-71-6

IUPAC Name: (2-amino-1-phenylpropyl)diphenylphosphane
Synonyms: (2-amino-1-phenylpropyl)diphenylphosphane
Molecular Weight (g/mol): 319.39
Molecular Formula: C21H22NP
InChi Key: JWZAIGGNEGTDMG-UHFFFAOYNA-N
SMILES: CC(N)C(P(C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results