CAS RN 342405-23-6
IUPAC Name:
2-(benzylamino)-1,3-thiazole-5-carboxylic acid
Synonyms:
2-(benzylamino)-1,3-thiazole-5-carboxylic acid
Molecular Weight (g/mol):
234.27
Molecular Formula:
C11H10N2O2S
InChi Key:
REVWBJBCAIBQGE-UHFFFAOYSA-N
SMILES:
OC(=O)C1=CN=C(NCC2=CC=CC=C2)S1
1
–
1
of
1
results
1
2-(Benzylamino)-1,3-thiazole-5-carboxylic acid, ≥95%, Thermo Scientific™
CAS: 342405-23-6 Molecular Formula: C11H10N2O2S Molecular Weight (g/mol): 234.27 MDL Number: MFCD03086200 InChI Key: REVWBJBCAIBQGE-UHFFFAOYSA-N Synonym: 2-benzylamino-1,3-thiazole-5-carboxylic acid,maybridge3_004319,2-benzylamino thiazole-5-carboxylic acid,5-thiazolecarboxylicacid, 2-phenylmethyl amino PubChem CID: 2779647 IUPAC Name: 2-(benzylamino)-1,3-thiazole-5-carboxylic acid SMILES: OC(=O)C1=CN=C(NCC2=CC=CC=C2)S1