accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 342405-38-3

IUPAC Name: 5-methyl-1-phenyl-1H-pyrazole-4-sulfonyl chloride
Synonyms: 5-methyl-1-phenylpyrazole-4-sulfonyl chloride
Molecular Weight (g/mol): 256.7
Molecular Formula: C10H9ClN2O2S
InChi Key: IRJCRNZTDRODFQ-UHFFFAOYSA-N
SMILES: CC1=C(C=NN1C1=CC=CC=C1)S(Cl)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results