accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 3425-46-5

KSeN

CAS RN 3425-46-5

IUPAC Name: potassium cyanoselanide
Synonyms: potassium selenocyanate
Molecular Weight (g/mol): 144.09
Molecular Formula: CKNSe
InChi Key: KYEKHFSRAXRJBR-UHFFFAOYSA-M
SMILES: [K+].[Se-]C#N
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)