accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HONHOO

CAS RN 3426-71-9

IUPAC Name: benzyl N-hydroxycarbamate
Synonyms: benzyl N-hydroxycarbamate
Molecular Weight (g/mol): 167.16
Molecular Formula: C8H9NO3
InChi Key: PQBSPTAPCMSZAA-UHFFFAOYSA-N
SMILES: ONC(=O)OCC1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)