accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 34301-54-7

SH

CAS RN 34301-54-7

IUPAC Name: adamantane-1-thiol
Synonyms: adamantane-1-thiol
Molecular Weight (g/mol): 168.3
Molecular Formula: C10H16S
InChi Key: ADJJLNODXLXTIH-UHFFFAOYSA-N
SMILES: SC12CC3CC(CC(C3)C1)C2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results