accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 34328-46-6

IUPAC Name: 4-chloro-3-(trifluoromethyl)benzaldehyde
Synonyms: 4-chloro-3-(trifluoromethyl)benzaldehyde
Molecular Weight (g/mol): 208.56
Molecular Formula: C8H4ClF3O
InChi Key: NIHMMULLFBKTOK-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC(C=O)=CC=C1Cl
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)