accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 343336-76-5

(S)(S)(R)(S)O(S)O(S)(S)(R)(R)OOHOHOHHOHOHOOHHO

CAS RN 343336-76-5

IUPAC Name: 2-(hydroxymethyl)-6-{[3,4,5-trihydroxy-5-(hydroxymethyl)oxolan-2-yl]methoxy}oxane-3,4,5-triol hydrate
Synonyms: melibiulose hydrate
Molecular Weight (g/mol): 360.31
Molecular Formula: C12H24O12
InChi Key: XZKUCJJNNDINKX-UHFFFAOYNA-N
SMILES: O.OCC1OC(OCC2OC(O)(CO)C(O)C2O)C(O)C(O)C1O
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results