accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 343880-24-0

CH3OOHO

CAS RN 343880-24-0

IUPAC Name: 2-[4-(methoxymethyl)phenyl]acetic acid
Synonyms: [4-(methoxymethyl)phenyl]acetic acid
Molecular Weight (g/mol): 180.2
Molecular Formula: C10H12O3
InChi Key: UBICBXUVKYZJTD-UHFFFAOYSA-N
SMILES: COCC1=CC=C(CC(O)=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results