accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 343926-69-2

NNBrH2N

CAS RN 343926-69-2

IUPAC Name: 4-bromopyrimidin-2-amine
Synonyms: 4-bromopyrimidin-2-amine
Molecular Weight (g/mol): 174
Molecular Formula: C4H4BrN3
InChi Key: MINURGGJJKMQQQ-UHFFFAOYSA-N
SMILES: NC1=NC=CC(Br)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results