accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

FFFClBr

CAS RN 344-65-0

IUPAC Name: 1-bromo-4-chloro-2-(trifluoromethyl)benzene
Synonyms: 1-bromo-4-chloro-2-(trifluoromethyl)benzene
Molecular Weight (g/mol): 259.45
Molecular Formula: C7H3BrClF3
InChi Key: OSTIALFVJOFNPP-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC(Cl)=CC=C1Br

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results