accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 34404-30-3

H3CH3CCH3OONHOHOOO

CAS RN 34404-30-3

IUPAC Name: 5-(benzyloxy)-4-{[(tert-butoxy)carbonyl]amino}-5-oxopentanoic acid
Synonyms: 5-(benzyloxy)-4-[(tert-butoxycarbonyl)amino]-5-oxopentanoic acid
Molecular Weight (g/mol): 337.37
Molecular Formula: C17H23NO6
InChi Key: CVZUKWBYQQYBTF-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NC(CCC(O)=O)C(=O)OCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results