accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

PPPdClClPht-Butylt-Butylt-Butylt-ButylPh

CAS RN 34409-44-4

IUPAC Name: bis(di-tert-butyl(phenyl)phosphane); dichloropalladium
Synonyms: bis(di-tert-butyl(phenyl)phosphane); palladium chloride
Molecular Weight (g/mol): 621.94
Molecular Formula: C28H46Cl2P2Pd
InChi Key: QNNRAWADYXIGHE-UHFFFAOYSA-L
SMILES: Cl[Pd]Cl.CC(C)(C)P(C1=CC=CC=C1)C(C)(C)C.CC(C)(C)P(C1=CC=CC=C1)C(C)(C)C
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results