accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CSO

CAS RN 3446-89-7

IUPAC Name: 4-(methylsulfanyl)benzaldehyde
Synonyms: 4-(methylsulfanyl)benzaldehyde
Molecular Weight (g/mol): 152.21
Molecular Formula: C8H8OS
InChi Key: QRVYABWJVXXOTN-UHFFFAOYSA-N
SMILES: CSC1=CC=C(C=O)C=C1
Molecular Weight (g/mol) 
  • (5)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (5)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)