accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CNCH3SFOO

CAS RN 34523-28-9

IUPAC Name: 5-(dimethylamino)naphthalene-1-sulfonyl fluoride
Synonyms: 5-(dimethylamino)naphthalene-1-sulfonyl fluoride
Molecular Weight (g/mol): 253.29
Molecular Formula: C12H12FNO2S
InChi Key: JMHHECQPPFEVMU-UHFFFAOYSA-N
SMILES: CN(C)C1=C2C=CC=C(C2=CC=C1)S(F)(=O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results