CAS RN 3453-00-7
IUPAC Name:
diethyl (2-oxo-2-phenylethyl)phosphonate
Synonyms:
diethyl 2-oxo-2-phenylethylphosphonate
Molecular Weight (g/mol):
256.24
Molecular Formula:
C12H17O4P
InChi Key:
HPEVTTNSIPGLEL-UHFFFAOYSA-N
SMILES:
CCOP(=O)(CC(=O)C1=CC=CC=C1)OCC