CAS RN 34552-15-3
IUPAC Name:
2-chloro-3-fluoro-5-methylpyridine
Synonyms:
2-chloro-3-fluoro-5-methylpyridine
Molecular Weight (g/mol):
145.56
Molecular Formula:
C6H5ClFN
InChi Key:
XZJURWKNRAGMCG-UHFFFAOYSA-N
SMILES:
CC1=CC(F)=C(Cl)N=C1
1
–
1
of
1
results
1
2-Chloro-3-fluoro-5-methylpyridine, 95%, Thermo Scientific Chemicals
CAS: 34552-15-3 Molecular Formula: C6H5ClFN Molecular Weight (g/mol): 145.56 MDL Number: MFCD06658238 InChI Key: XZJURWKNRAGMCG-UHFFFAOYSA-N Synonym: 2-chloro-3-fluoro-5-picoline,2-chloro-3-fluoro-5-methyl-pyridine,6-chloro-5-fluoro-3-picoline,pyridine, 2-chloro-3-fluoro-5-methyl,2-chloro-3-fluoro-5-methyl pyridine,pubchem14231,ksc495q2t,6-chloro-5-fluoro-3-methylpyridine PubChem CID: 13541365 IUPAC Name: 2-chloro-3-fluoro-5-methylpyridine SMILES: CC1=CC(F)=C(Cl)N=C1