accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3H3CMgBr

CAS RN 34589-46-3

IUPAC Name: magnesium(2+) 2,4-dimethylbenzen-1-ide bromide
Synonyms: magnesium(2+) 2,4-dimethylbenzen-1-ide bromide
Molecular Weight (g/mol): 209.37
Molecular Formula: C8H9BrMg
InChi Key: ATHZRUUXJPXJNP-UHFFFAOYSA-M
SMILES: [Mg++].[Br-].CC1=CC=[C-]C(C)=C1
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)