accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 345909-26-4

IUPAC Name: 2-(4-{4,7,11-trihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl}pentanamido)ethane-1-sulfonic acid sodium
Synonyms: taurocholic acid sodium
Molecular Weight (g/mol): 538.7
Molecular Formula: C26H45NNaO7S
InChi Key: LHLYEKPSOJJBMK-UHFFFAOYNA-N
SMILES: [Na].CC(CCC(=O)NCCS(O)(=O)=O)C1CCC2C3C(O)CC4CC(O)CCC4(C)C3CC(O)C12C
Molecular Weight (g/mol) 
  • (3)
  • (4)
  • (3)
Quantity 
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
Percent Purity 
  • (4)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results