accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHFFF

CAS RN 346-06-5

IUPAC Name: [2-(trifluoromethyl)phenyl]methanol
Synonyms: [2-(trifluoromethyl)phenyl]methanol
Molecular Weight (g/mol): 176.14
Molecular Formula: C8H7F3O
InChi Key: TWQNSHZTQSLJEE-UHFFFAOYSA-N
SMILES: OCC1=CC=CC=C1C(F)(F)F
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (5)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)