accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 3460-24-0

BrClBr

CAS RN 3460-24-0

IUPAC Name: 1,4-dibromo-2-chlorobenzene
Synonyms: 1,4-dibromo-2-chlorobenzene
Molecular Weight (g/mol): 270.35
Molecular Formula: C6H3Br2Cl
InChi Key: LOWQAATYMJIWOG-UHFFFAOYSA-N
SMILES: ClC1=C(Br)C=CC(Br)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results