accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 3466-32-8

IUPAC Name: 1-bromo-4-methanesulfonylbenzene
Synonyms: 1-bromo-4-methanesulfonylbenzene
Molecular Weight (g/mol): 235.1
Molecular Formula: C7H7BrO2S
InChi Key: FJLFSYRGFJDJMQ-UHFFFAOYSA-N
SMILES: CS(=O)(=O)C1=CC=C(Br)C=C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)