accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NFFF

CAS RN 347-42-2

IUPAC Name: 2-(trifluoromethyl)quinoline
Synonyms: 2-(trifluoromethyl)quinoline
Molecular Weight (g/mol): 197.16
Molecular Formula: C10H6F3N
InChi Key: YZSRICFIQLVSMQ-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=NC2=CC=CC=C2C=C1
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)