accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHO

CAS RN 3470-49-3

IUPAC Name: 5-hydroxy-2,3-dihydro-1H-inden-1-one
Synonyms: 5-hydroxy-2,3-dihydroinden-1-one
Molecular Weight (g/mol): 148.16
Molecular Formula: C9H8O2
InChi Key: ZRKQOVXGDIZYDS-UHFFFAOYSA-N
SMILES: OC1=CC=C2C(=O)CCC2=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)