accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 3473-63-0

IUPAC Name: acetic acid; methanimidamide
Synonyms: acetic acid; formamidine
Molecular Weight (g/mol): 104.11
Molecular Formula: C3H8N2O2
InChi Key: XPOLVIIHTDKJRY-UHFFFAOYSA-N
SMILES: NC=N.CC(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results