accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 348-40-3

IUPAC Name: 6-fluoro-1,3-benzothiazol-2-amine
Synonyms: 6-fluoro-1,3-benzothiazol-2-amine
Molecular Weight (g/mol): 168.19
Molecular Formula: C7H5FN2S
InChi Key: CJLUXPZQUXVJNF-UHFFFAOYSA-N
SMILES: NC1=NC2=CC=C(F)C=C2S1
Molecular Weight (g/mol) 
  • (4)
  • (3)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (4)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (4)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (2)