accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHClF

CAS RN 348-60-7

IUPAC Name: 4-chloro-3-fluorophenol
Synonyms: 4-chloro-3-fluorophenol
Molecular Weight (g/mol): 146.55
Molecular Formula: C6H4ClFO
InChi Key: XLHYAEBESNFTCA-UHFFFAOYSA-N
SMILES: OC1=CC=C(Cl)C(F)=C1
Boiling Point 
  • (1)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)