accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

(S)ONH2OOCH3HCl

CAS RN 34805-17-9

IUPAC Name: methyl 2-amino-3-[4-(benzyloxy)phenyl]propanoate hydrochloride
Synonyms: methyl 2-amino-3-[4-(benzyloxy)phenyl]propanoate hydrochloride
Molecular Weight (g/mol): 321.8
Molecular Formula: C17H20ClNO3
InChi Key: IQKXGACPIPNLEL-UHFFFAOYNA-N
SMILES: Cl.COC(=O)C(N)CC1=CC=C(OCC2=CC=CC=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results