accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 34841-11-7

IUPAC Name: methyl 2-methoxy-5-nitrobenzoate
Synonyms: methyl 2-methoxy-5-nitrobenzoate
Molecular Weight (g/mol): 211.17
Molecular Formula: C9H9NO5
InChi Key: DOBFJVVTBNTGCW-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(OC)C=CC(=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results