accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 34893-92-0

IUPAC Name: 1,3-dichloro-5-isocyanatobenzene
Synonyms: 1,3-dichloro-5-isocyanatobenzene
Molecular Weight (g/mol): 188.01
Molecular Formula: C7H3Cl2NO
InChi Key: XEFUJGURFLOFAN-UHFFFAOYSA-N
SMILES: ClC1=CC(=CC(Cl)=C1)N=C=O
Molecular Weight (g/mol) 
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results