accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 349-57-5

NOOHOFFF

CAS RN 349-57-5

IUPAC Name: 3-nitro-5-(trifluoromethyl)phenol
Synonyms: 3-nitro-5-(trifluoromethyl)phenol
Molecular Weight (g/mol): 207.11
Molecular Formula: C7H4F3NO3
InChi Key: JDIAMHNYAPDMRB-UHFFFAOYSA-N
SMILES: OC1=CC(=CC(=C1)[N+]([O-])=O)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results