accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 349-58-6

OHFFFFFF

CAS RN 349-58-6

IUPAC Name: 3,5-bis(trifluoromethyl)phenol
Synonyms: 3,5-bis(trifluoromethyl)phenol
Molecular Weight (g/mol): 230.11
Molecular Formula: C8H4F6O
InChi Key: ODSXJQYJADZFJX-UHFFFAOYSA-N
SMILES: OC1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)