accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CNHOCH3OH3C

CAS RN 3490-06-0

IUPAC Name: [2-(3,4-dimethoxyphenyl)ethyl](methyl)amine
Synonyms: [2-(3,4-dimethoxyphenyl)ethyl](methyl)amine
Molecular Weight (g/mol): 195.26
Molecular Formula: C11H17NO2
InChi Key: HNJWKRMESUMDQE-UHFFFAOYSA-N
SMILES: CNCCC1=CC=C(OC)C(OC)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results