accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CCH3CH3NONOONOO

CAS RN 349091-80-1

IUPAC Name: N-benzyl-N-tert-butyl-3,5-dinitrobenzamide
Synonyms: N-benzyl-N-tert-butyl-3,5-dinitrobenzamide
Molecular Weight (g/mol): 357.37
Molecular Formula: C18H19N3O5
InChi Key: OLMWJDGSOFEXIR-UHFFFAOYSA-N
SMILES: CC(C)(C)N(CC1=CC=CC=C1)C(=O)C1=CC(=CC(=C1)[N+]([O-])=O)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results