accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 3491-63-2

IUPAC Name: (2E)-2-phenylpent-2-enal
Synonyms: (2E)-2-phenylpent-2-enal
Molecular Weight (g/mol): 160.22
Molecular Formula: C11H12O
InChi Key: YPAJRUMMODCONM-WDZFZDKYSA-N
SMILES: CC\C=C(\C=O)C1=CC=CC=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results