accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 349396-05-0

CH3NONOO

CAS RN 349396-05-0

IUPAC Name: N-benzyl-N-ethyl-4-nitrobenzamide
Synonyms: N-benzyl-N-ethyl-4-nitrobenzamide
Molecular Weight (g/mol): 284.32
Molecular Formula: C16H16N2O3
InChi Key: MWXUHJACDKHYRK-UHFFFAOYSA-N
SMILES: CCN(CC1=CC=CC=C1)C(=O)C1=CC=C(C=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results