accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 35013-72-0

IUPAC Name: 2,5-dioxopyrrolidin-1-yl 5-{2-oxo-hexahydro-1H-thieno[3,4-d]imidazol-4-yl}pentanoate
Synonyms: 2,5-dioxopyrrolidin-1-yl 5-{2-oxo-hexahydrothieno[3,4-d]imidazol-4-yl}pentanoate
Molecular Weight (g/mol): 341.38
Molecular Formula: C14H19N3O5S
InChi Key: YMXHPSHLTSZXKH-UHFFFAOYNA-N
SMILES: O=C(CCCCC1SCC2NC(=O)NC12)ON1C(=O)CCC1=O
Molecular Weight (g/mol) 
  • (9)
  • (3)
  • (2)
Quantity 
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
Percent Purity 
  • (6)
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (3)
  • (3)
  • (6)
  • (9)
  • (3)
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (6)
  • (5)
  • (5)