accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CBr

CAS RN 3508-00-7

IUPAC Name: 1-bromoheptadecane
Synonyms: 1-bromoheptadecane
Molecular Weight (g/mol): 319.37
Molecular Formula: C17H35Br
InChi Key: HHSDZLLPIXMEIU-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCBr
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results