accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3SH2NOOCH3

CAS RN 350988-88-4

IUPAC Name: methyl 2-amino-5-methyl-4-phenylthiophene-3-carboxylate
Synonyms: methyl 2-amino-5-methyl-4-phenylthiophene-3-carboxylate
Molecular Weight (g/mol): 247.31
Molecular Formula: C13H13NO2S
InChi Key: AIHYNLWSPTZXGT-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(N)SC(C)=C1C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results