accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHOFFF

CAS RN 351-35-9

IUPAC Name: 2-[3-(trifluoromethyl)phenyl]acetic acid
Synonyms: [3-(trifluoromethyl)phenyl]acetic acid
Molecular Weight (g/mol): 204.15
Molecular Formula: C9H7F3O2
InChi Key: BLXXCCIBGGBDHI-UHFFFAOYSA-N
SMILES: OC(=O)CC1=CC=CC(=C1)C(F)(F)F
Molecular Weight (g/mol) 
  • (2)
  • (6)
Percent Purity 
  • (2)
  • (6)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (2)
  • (6)
  • (2)
  • (6)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)