accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ONOClOHO

CAS RN 351003-03-7

IUPAC Name: 3-(6-chloro-3-oxo-3,4-dihydro-2H-1,4-benzoxazin-4-yl)propanoic acid
Synonyms: 3-(6-chloro-3-oxo-2H-1,4-benzoxazin-4-yl)propanoic acid
Molecular Weight (g/mol): 255.65
Molecular Formula: C11H10ClNO4
InChi Key: REQQIWCFXWGLDY-UHFFFAOYSA-N
SMILES: OC(=O)CCN1C(=O)COC2=C1C=C(Cl)C=C2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results